Description
- Minimized plumbing connections
- Integrated full-flow fluid strainer
- Four pump options
- Supports applications requiring temperatures of -10 °C to 30 °C
Specifications
5 gal. | |
5.3 L/min. | |
RS232 - digital communication optional (located on the back of the unit) | |
208/230 V 60 Hz |
19 L | |
PD1 (Positive Displacement Pump) | |
28.6 x 17.3 x 23.6 in (72.6 x 43.9 x 59.9 cm) |