Learn More
Leica Microsystems ThermoBrite™ Slide Denaturization and Hybridization System
Automates pre- and post hybridization steps in FISH testing
Supplier: Leica Microsystems 3800004852002
Description
- Provides on-board denaturation and hybridization
- Can process 12 or 24 slides per run
- Just load slides and walk away
- Temperature control: +1°C
- Easy to use, intuitive software allows user to run standard protocols for blood, urine or solid tumor specimens
- Can be fit virtually anywhere in laboratory due to its small size
- Provides consistency and reproducibility and help save time and money
Specifications
45.1 x 22.9 x 13.5 cm (17.75 x 9 x 5.5 in.) | |
240 V | |
0 to 99 hrs, Continuous Mode; 30°C to 99°C |
24 Slides per run | |
Pre- and Post Hybridization Steps in FISH Testing |