Learn More
Unity Lab Services™ Thermo Scientific™ Ultra-Low Temperature Freezers Validation Documention
As an original equipment manufacturer (OEM) service provider for Thermo Fisher Scientific, all protocols designed to optimize efficiency are included with each Thermo Scientific Ultra-Low Temperature Freezers Models: Thermo Scientific TDE, Forma FDE, Revco RDE, Isotemp IUE
Supplier: Unity Lab Services™ IOQDOCE89003484
Description
IQ/OQ documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept up to date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Thermo Scientific Ultra-Low Temperature Freezers including all Thermo Scientific Ultra-Low Temperature Freezers Models: Thermo Scientific TDE, Forma FDE, Revco RDE, Isotemp IUE | |
Documentation Only for the following, Services Not Included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform empty chamber temperature mapping, open door and power loss testing. | |
Contact your local sales representative for a list of standard and extended coverage areas. | |
IQ/OQ Documentation |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.