Learn More
Thermo Scientific™ TSX Series High-Performance Lab Refrigerators
Innovative refrigeration solutions for healthcare and research needs.
Supplier: Thermo Scientific™ TSX1205SV
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
- Designed to safely store medical and laboratory products, including vaccines, pharmaceuticals, media and reagents requiring a temperature range of 2°C to 8°C
- Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
- Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
- Up to 40% less energy usage than conventional refrigerant models*
- Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP)
- Manufactured in an award-winning, zero-waste to landfill facility
- Operating temperature range of 2° to 8°C, factory setpoint at 5°C
- Choice of glass or solid door models
- Lower HVAC costs as a result of less heat emission into the environment*
- Easy to use microprocessor control with interior light, setpoint security and alarms
- Compatible with wireless monitoring systems and remote alarm contacts
- Standard door locks provide comfort and assurance
- Ergonomic door handle
- Optional chart recorders available
- Warranty varies by country, please contact us for details
Wireless Monitoring Solution
Thermo Scientific™ DeviceLink HUB™, our easy-to-install solution, works with the Thermo Fisher Cloud to monitor conditions inside your TSX Series Laboratory Refrigerators and Freezers.
DeviceLink HUB™ connects users to their equipment, allowing them to receive critical lab equipment data and equipment notifications on their mobile and web applications:
- Display temperature
- Ambient Temperature
- Alarm and alerts notification
DeviceLink HUB™ is available in the following regions:
- United States and Canada
- 28 members of the European Union
- Iceland, Liechtenstein, Norway, and Switzerland
Specifications
High-Performance Refrigerator | |
326 L | |
R290 | |
1 | |
UL, cUL, CE | |
TSX Series | |
Alarm, Recorder | |
Smart-Vue Compatible | |
21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm) | |
31.1 in. | |
79.0 cm | |
73.0 in. | |
185.4 cm | |
24.0 in. | |
61.9 cm | |
15 A | |
230 V | |
50/60 Hz | |
154 kg | |
130 kg |
11.5 cu. ft. | |
3°-7°C (5°C) | |
Solid | |
4 | |
Microprocessor | |
Automatic | |
Optional | |
31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm) | |
31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm) | |
21.8 in. | |
55.4 cm | |
52.4 in. | |
133.1 cm | |
20 in. | |
50.8 cm | |
CEE 7/7 | |
208-230V, 50Hz | |
340 lb. | |
286 lb. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.