Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ SUN-SRi™ Standard Opening/StepVial II™ Inserts
SUN-Sri Inserts are available in a variety of configurations to accommodate small sample size.
Catalog Number | Mfr. No. | Material | Style | Volume (English) Insert | Volume (Metric) Insert | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Style | Volume (English) Insert | Volume (Metric) Insert | Price | Quantity & Availability | ||
22-021894
|
Thermo Scientific™
200 232 |
Glass | Flat | 0.006 oz. | 0.2 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||
22-021893
|
Thermo Scientific™
200 228 |
Glass | Flat | 0.006 oz. | 0.2 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||
22-021895
|
Thermo Scientific™
200 238 |
Glass | Conical | 0.006 oz. | 0.2 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||
14-823-327
|
Thermo Scientific™
500 208 |
Glass | Conical | 0.006 oz. | 200 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
22-035113
|
Thermo Scientific™
500 918 |
Borosilicate Glass (Type 1 Class A) | Mandrel | 0.003 oz. | 0.1 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||
14-823-386
|
Thermo Scientific™
501 330 |
Polypropylene | Conical | 0.006 oz. | 200 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
Description
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title