Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ SUN-SRi™ Septa for 8-425 Screw Thread Caps
Thermo Scientific caps and vials are available in a wide variety of sizes and configurations to fit the needs of many applications.
Supplier: Thermo Scientific™ 500 870
Description
Specifications
0.314 in. | |
0.8 cm | |
Yes | |
Not Autoclavable |
Red, White | |
Red PTFE, Silicone | |
0.045 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction