Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ SUN-SRi™ GC Septa
Designed to operate at true temperatures of 290° or 306°C
Supplier: Thermo Scientific™ 502 289
Description
Chromseal 9001 High Temperature GC
- Designed to perate at true temperatures of 306°C.
- Red septa
- Designed to operate at a true temperature of 290°C
- Offer a longer operational life at recommended temperature
- Blue septa
Order Info
are available in appropriate diameters to fit a variety of gas chromatographs.
Specifications
Septa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction