Learn More
Epredia™ Shandon™ Electronic Floor Scale
Crafted from epoxy-painted mild steel
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
20565
|
Epredia™
20565 |
Quick Pit Frame | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
20560
|
Epredia™
20560/DEL |
Floor Scale | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | |||||
20570
|
Epredia™
20570/DEL |
Digital Indicator | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | |||||
Description
- A complete line of of epoxy-painted mild steel, electronic floor scales are available
- Variety of sizes are available from 3 × 3 ft. (91.4 × 91.4cm) to 5 × 7 ft. (152.4 × 213.4cm)
- Ramped models are also available
- Variety of capacities are available from 1000 to 10,000 lb. (453.6 to 4536kg)
- No mechanical adjustment is required
- Hermetically sealed load-sensing elements allow the user to clean the unit without damaging the weighing mechanisms
- Standard scale is installed into a pit, but ramped and freestanding units are available
- Pimarily constructed of stainless steel, including the load cells, weighbridge, scale frame, optional ramps
Order Info
Scale 20560 does not include pit frame, ramp, or digital indicator (sold separately)
Certifications
A Certificate of Conformance, Number 91-097 was issued under the National Type Evaluation Program (NTEP).
Compliance
Scales meet /exceed the Class III, 5000 division accuracy with the National Institute of Standards and Technology (NIST™) Handbook 44.