Learn More
Thermo Scientific™ QCC (Quick Column Change) for Agilent Instruments
This Quick Column Change accessory is compatible with Agilent Instrumentation
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
60180872
|
Thermo Scientific™
60180872 |
S/SL Injector Adaptor | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
60180878
|
Thermo Scientific™
60180878 |
TCD Detector Adaptor | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
60180873
|
Thermo Scientific™
60180873 |
Tight Nut | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
QCC (Quick Column Change)-Leak Free Column Installation
- Tool-free installation of analytical GC column
- Graphite ferrules
Complete QCC Kits Includes:
- Injector Adaptor
- Detector Adaptor
- Tight Nut
- Ferrules (1 each 0.25, 0.32,0.53)
- Ceramic Column Cutter