Learn More
Thermo Scientific™ Orion Star™ A211 pH Benchtop Meter Promotion
Reliably measure pH, mV, ORP and temperature with the Thermo Scientific™ Orion™ Star A211 pH Benchtop Meter for advanced lab analysis and printer or computer interfacing.
Supplier: Thermo Scientific™ STARA2115PM
Description
Record accurate and reliable pH, mV, ORP and temperature measurements with the Thermo Scientific™ Orion™ Star A211 pH Benchtop Meter, ideal for a wide range of applications and advanced pH analysis in the lab. Perform up to a five point pH calibration, log up to 2000 data point sets with time / date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer with free downloadable Orion™ Star Com software. Use the included electrode stand to easily place sensors in samples and simplify storage to minimize breakage.
- Plain language screen prompts, soft keys that update for easy selection
- Multilanguage interface that supports English, Spanish, French, Italian, German and Chinese
- AUTO-READ™ that locks in the stable reading on your screen, ready indicator alerts when readings are stable and timed reading gathers data in specific time intervals
- Up to five point pH calibration with automatic recognition for USA/NIST™ and DIN buffers
- Calibration editing for fixing calibration errors without a complete recalibration
- Non-volatile memory to hold up to 2000 data points with time and date stamp
- USB and RS232 ports and complimentary data analysis software to easily transfer data and keep meter software up-to-date
- Direct control of the Orion Star stirrer probe (096019, sold separately) for mixing samples without a stir plate
- Electrode arm and newly designed probe holder make it easier to maintain and place probes into samples
- Universal power adapter works with almost every AC power source, or use four AA batteries (sold separately) to run the meter on DC power
- IP54-rated housing handles splashes
- Wall-mountable if table space is a problem
Specifications
Orion Star A211 pH benchtop meter kit with ROSS Ultra Triode glass-body pH/ATC electrode, solutions and stand | |
±0.2 or ±0.05%, whichever is greater | |
Transfer single, range or all data points to printer or computer | |
Star A211 pH meter, 8302BNUMD ROSS Ultra Triode glass-body pH/ATC electrode, 810199 ROSS pH buffer and solution kit, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate | |
pH with calibration editing option, relative mV, ORP and temperature | |
0.001, 0.01, 0.1 | |
±2000.0mV | |
1 to 5 | |
22.0°F to 221.0°F | |
Manual or automatic with ATC temperature probe | |
LCD, Graphic with Backlight | |
Non-volatile memory preserves data log, calibration log and meter settings | |
Automatic with Auto-Read and Timed Read Modes, Manual with Continuous Read Mode | |
BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe) | |
IP-54 | |
24 cm x 18 cm x 11 cm (9.5 in. x 7.1 in. x 4.3 in.) | |
2 lb. | |
Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys | |
Orion Star A211 pH/mV/RmV/ORP/Temperature Benchtop Meter | |
pH electrode, ORP electrode, reference half-cell electrode, ATC temperature probe, stirrer probe (includes 8302BNUMD ROSS Ultra Triode pH/ATC electrode) | |
±2000.0mV | |
7.000pH | |
41°F to 113°F (ambient) | |
1 | |
800 hr. |
±0.1°C | |
±0.002 | |
USB, RS-232 | |
pH, mV, Relative mV, ORP with Temperature | |
-2.000 to 20.000 | |
±2000 | |
0.1mV | |
-5.0°C to +105.0°C | |
0.1°C, 0.1°F | |
1 point temperature offset calibration | |
Yes, selectable | |
2000 with Date and Time Stamp | |
High/low limit alarm, calibration due alarm | |
CE, TUV 3-1, FCC Class A | |
Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003) | |
0.9 kg | |
3 Year Meter Warranty | |
10 per channel with password protection | |
Universal AC adapter (included) or 4 AA batteries (sold separately) | |
English, Spanish, French, Italian, German and Chinese; Portuguese and Korean Available with Software Update | |
5 to 85%, non-condensing | |
Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting | |
5°C to 45°C (ambient) | |
4 x AA (sold separately) |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.