Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Orion™ Green pH Combination Electrodes
Available in low maintenance gel versions for simplified preparation and use or refillable version for improved longevity.
Supplier: Thermo Scientific™ GS9106BNWP/EMD
Description
- RoHS compliant—no lead, mercury or other hazardous materials
- No disposal issues, no disposal cost
- Choice of single-junction models for routine sample measurements, or double-junction models for biological samples or samples that contain Tris or sulfides
- Choice of refillable or low-maintenance gel-filled models
- Duarable epoxy body
- pH Range: 0 to 14
- Dimensions: 120 x 12mm, cap dia. 16mm
- Waterproof BNC connector, 1m cable; fit any meter with BNC input
Specifications
WPBNC | |
Epoxy Body | |
0°C to 80°C | |
1 m |
Single | |
0 to 14 | |
39.3 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction