Learn More
Thermo Scientific™ Nunc™ Strips and Frames
Ideal for sequencing, restriction analysis, transformations, ligations and storage
Supplier: Thermo Scientific™ 232034/PRC
Description
Tape and well design reduce evaporation.
- Centrifugation eliminated when mixing reagents
- Easy well identification
- Minimum binding of hydrophilic molecules
- Standard 96 well format
- Modules are temperature resistant from -70°C to +121°C
- Space saving at the bench and in the freezer
- Plastic waste is reduced
Specifications
Strip Plate | |
8 x 12 | |
Polypropylene | |
96 |