Learn More
Epredia™ Lab Vision™ Autostainer 720-2D
Handles up to 84 slides at once, making it the ideal solution for high-throughput laboratories
Supplier: Epredia™ A80500030
Includes: Autostainer 720 2D System: 84-slide Autostainer with 2D matrix bar code reader; Computer; Monitor; Label printer(s); Document printer; Seven metal reinforced, ABS plastic slide racks; Reagent rack; Reagent vials and caps; Autostainer probe cleaning reagent kit; Buffer rinse bottle; Wash Buffer, DI water, and waste containers; Universal Power Supply.
Description
- Up to 84-slide capacity, each individually programmable
- User-specified protocols and choice of reagents
- Multiple staining runs per day
- Truly hands-free, walk-away operations with advanced timer functions for overnight use
- Easy, Windows™ 7 Professional-based programming
- Modular options with up to four units controlled from one PC
- Complemented by the Epredia PT Module designed for fully automated, simultaneous dewaxing and epitope recovery (PT Module also sold separately)
System Features
Incorporates 2D Data Matrix Bar Coding for Slides and Reagents
- High precision label detector technology for rapid scanning
- Integrated digital bar code scanner
- Fully configured 2D data matrix software and printer interface
- Bar code control of all protocol, reagent, and slide operations
- Preprintable data matrix labels with user-defined Autoprograms
- Adaptable for proprietary 2D-labeled reagents, or user-defined reagents/labels
- Optional, individual inactivation of slide and/or reagent bar coding