Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ GENESYS™ 20 Spectrophotometer Test Tube Holder for 1 in. Tubes
Manufacturer: Thermo Scientific™ 4015000
Description
“Snap-in” holder fits 1 in. test tubes up to 104mm tall.
Specifications
Test Tube Holder | |
Genesys™ 20 spectrophotometer | |
One test tube | |
104 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title