Learn More
Fisherbrand™ Flexaframe™ End-to-End Rod Connector
Strong aluminum alloy connector permits end-to-end joining of rods
This item is not returnable.
View return policy
Description
- Extends length of Flexaframe
- Precision boring ensures perfect aligning of rods
- Corrosion resistant
- 2L x 0.87 in. O.D. (51 × 22mm) with two setscrews
Specifications
Aluminum Alloy | |
5.1 cm | |
2.2 cm | |
End to End Connector |
2 in. | |
0.87 in. | |
Two setscrews | |
Permits end-to-end joining of rods, precision boring ensures perfect aligning of rods |