Learn More
Unity Lab Services™ Thermo Scientific™ Centrifugation Validation Service with Cycle Testing
Stay compliant with our Industry-Exclusive, No-Charge Requalification Guarantee included with each Thermo Scientific Centrifugation Validation Service with Cycle Testing for Benchtop, Micro, Bioprocessing, Clinical, and Standard floor model centrifuges: Thermo Scientific, Sorvall
Supplier: Unity Lab Services™ IOPQPCKE89003492
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
This item is not returnable.
View return policy
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Thermo Scientific Centrifugation including Benchtop, Micro, Bioprocessing, Clinical, and Standard floormodel centrifuges: Thermo Scientific, Sorvall | |
Industry-Exclusive, No-Charge Requalification Guarantee. Unity Lab Services is the only OEM service provider to offer a guarantee and promise to requalify instruments and equipment at no charge if a key component fails while under the original factory warranty or a qualifying instrument service plan. | |
Travel to major metropolitan areas. Does not include Installation. Instrument must be installed, powered-up and set temperature point for 24 hours before on-site service |
Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform min/max speed, brief/extended time and temperature testing (for refrigerated models). Cycle Testing: Point-In-Time test exercise of pre-defined duration that surpasses singular calibration and routine speed, time, and temperature monitoring activities. | |
Contact your local sales representative for a list of standard and extended coverage areas. |