Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Bevel Tip Pipet Tips

Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip
Description
- Certified by independent laboratory
- Nonsterile: RNase, DNase, DNA, and pyrogen free
- Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and pyrogen free
- For use with most 200 and 1000μL single and multichannel pipetters
- Hinged racks can be stacked rack-on-rack
- Hinged closure on racks allows top lid to be partially open for steam penetration during autoclaving
Order Info
Shipped in racks, 960/Pack
Specifications
1 to 200 μL | |
Yellow | |
2 in. | |
DNase, RNase, DNA, and Pyrogen Free | |
Rack | |
Universal Fit Pipettor |
Non-sterile | |
Autoclavable | |
5cm | |
Universal Fit Pipettor | |
Beveled | |
PP (Polypropylene) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title