Learn More
Thermo Scientific™ ART™ Solvent Safe Carbon Filtered Pipette Tips
For acids, bases and aggressive organic solvents
$269.00
Specifications
Filtered | Filtered |
---|---|
Sterility | Non-sterile |
Tip Style | Universal Fit |
Product Line | ARTTM |
Material | Carbon with Polyethylene Filter |
Catalog Number | Mfr. No. | Volume (Metric) | Tip Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Tip Style | Price | Quantity & Availability | ||||
21-236-19C
|
Thermo Scientific™
5079EB |
100 μL to 1000 μL | Universal Fit |
Pack of 500 for $269.00
Case of 5 PK for $1,208.00 |
|
||||
Description
For acids, bases and aggressive organic solvents
Revolutionary filter material handles the pipetting rigors of Combinatorial Chemistry. Unique use of the ART™ filter with Folded Activated Carbon offers both pipetter and sample protection against destructive carryover aerosols and vapors from volatile organic solvents, all without compromising pipetting accuracy and precision.
- For manual pipetting of organic solvents
- Utilizes new formula of the ART (aerosol resistant tip) filter tip
- Helps to eliminate pipetting failures and critical inaccuracies caused by strong acids, bases and aggressive organic solvents
- Proprietary blend of Folded Activated Carbon with a 40-micron polyethylene filter provides effective solvent protection
Specifications
Filtered | |
Universal Fit | |
Carbon with Polyethylene Filter |
Non-sterile | |
ARTTM |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.