Learn More
Thermo Scientific™ Accessories for RadEye™ Radiation Detectors
Accessory for use with RadEye pocket-sized radiation detectors
$78.00 - $757.00
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
19-172-509
|
Thermo Scientific™
425067065 |
Docking Station |
Each for $757.00
|
|
|||||
19-172-510
|
Thermo Scientific™
425067064 |
Goose Neck Mounting Kit |
Each for $78.00
|
|
|||||
19-172-511
|
Thermo Scientific™
425067042 |
Headset |
Each for $110.00
|
|
|||||
Description
- Docking Station/Car Adapter includes charging circuitry (8-30 V DC, cigarette lighter plug), alarm relay, RS 232 interface, AAA NiMH rechargeable batteries
- Goose neck mounting kit has suction cup for instanteneous attachment to windshield; fits to docking station (19-172-509) or desktop holder
- Headset for RadEye includes earphone for RadEye series