Learn More
Cliawaived Inc Human Body Temperature Digital Non Contact Infrared Thermometer
Supplier: Cliawaived Inc CLIATSJXB178
(1 Thermometer)-Non-contact clinical thermometer. Measurement of body / object / room temperature.-Hygienic and safe-measure without skin contact.-Measurement in <1 second-Infrared measurement technology-Display in Centigrade or Fahrenheit-Free high temperature alarm setting-Batteries not included, requires 2 AA batteries.NOTE: Accuracy of Thermal Scanners maybe .5-1 degree lower than oral, while ear thermometers maybe + .5-1 higher. (Source Kaiser Permanente - https://wa.kaiserpermanente.org/kbase/topic.jhtml?docId=tw9223)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.