Learn More
Science Company THERMAL INSULATION WRAP
SDP
Supplier:  Science Company NC14136
Encompass
Thermal wrap your distillation column to prevent heat loss and increase the efficiency. Made from high temperature resistant material.
Catalog No. NC2059836