Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Thermal Conductivity Bars
SDPSCI_ED

Thermal Conductivity Bars quickly show heat transfer rates.

Supplier:  United Scientific Supplies CBR001

Catalog No. S82876


Add to Cart

Alert:

Prop 65 Warning

This item is not returnable. View return policy

Description

Description

  • This simple and ingenious device quickly and graphically shows students how different metals conduct heat at different rates.
  • It consists of four identically sized bars of common metals - steel, brass, aluminum, and copper - mounted parallel to one another in a plastic support.
  • Attached to each bar is a liquid crystal thermometer.
  • When the ends of the bars are immersed in hot water, the thermometers quickly show the progress of heat along each bar.
  • Students can measure the time for the tops of the bars to reach the top display temperature (42°C) and discover how well each metal conducts heat.
  • Height: 20 cm, Width: 5.5 cm, Weight: 145 g.
  • Bar dimensions (each bar): Length: 13.5 cm, Width: 0.9 cm, Thickness: 0.5 cm.
  • Display temperature range: 35°C - 42°C x 1°C.
Specifications

Specifications

5+
Conductivity Bar
145 g
8 in.
5 oz.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit