Learn More
Bachem Fmoc-Pen(Trt)-OH
Supplier: Bachem 4019778.0005
Fmoc-Pen(Trt)-OH, 201531-88-6, C39H35NO4S, Mr 613.78, 5g. Fmoc-b,b-dimethyl-Cys(Trt)-OH. The coupling of peptide thioesters to N-terminal penicillamine followed by desulfuration yielding valine, which was described by Haase et al., broadens the scope of native chemical ligation.