Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSI, LLC: The Case of the Kidnapped Cookies Kit
Who stole the cookies from the cookie jar?
Supplier: Aqua Phoenix Scientific CRI5303REFILL
Includes:
teacher's guide, student worksheets and casebooks and a rubric
Description
- Activity involves investigating the theft of Otis Spunkmeyer cookies
- Students need to figure out which of their staff members took the warm, chewy cookies
- Clues involve a ransom note written in Spanish, cut from several different local newspapers, with red stain (possibly lipstick?) and white powder and soil left near the note
- Students to determine whose DNA is on the envelope the note was found in
- Activity also involves trial for culprit
- Guilty party pays for free cookies for everyone as sentence
- For middle school students
- Class size: 30 students
- Requires a microscope (sold separately)
Specifications
the Case of the Kidnapped Cookies Lab Activity Refill | |
For the case of the kidnapped cookies lab activity kit | |
Microscope | |
30 Students |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction