Learn More
Perkin Elmer US LLC TFE Tracey Spray Chamber for Avio 200/500
Supplier: Perkin Elmer US LLC N0790373
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The 50 mL cyclonic spray chamber, tetrafluoroethylene (TFE), is a hydrofluoric acid-resistant spray chamber. It is o-ring-free.