Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Texwipe™ TexWrite™ Ring Binder
Supplier: Texwipe™ TX5810
Description
The TexWrite 1 in. (25mm) three-ring binder designed for cleanroom applications.
- Durable polyethylene cover and 100% plastic ring mechanism ensure cleanliness and corrosion resistance
- All rings open quickly with an easy-slide tab for adding or removing pages
Order Info
Each binder packaged in two polypropylene bags
Specifications
Latex-impregnated Paper | |
1 in. | |
Three Ring Binder |
Polyethylene | |
Document storage, equipment manuals, procedure, note taking |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction