Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylene glutarimide, 99%, Thermo Scientific™

Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1075-89-4
C9H13NO2
167.21
MFCD00023871
YRTHJMQKDCXPAY-UHFFFAOYSA-N
3,3-tetramethyleneglutarimide, 8-azaspiro 4.5 decane-7,9-dione, azaspirodecanedione, 1,1-cyclopentanediacetimide, 8-aza-spiro 4.5 decane-7,9-dione, buspirone impurity k, dsstox_cid_31405, dsstox_rid_97291, dsstox_gsid_57616
136843
8-azaspiro[4.5]decane-7,9-dione
C1CCC2(C1)CC(=O)NC(=O)C2
1075-89-4
C9H13NO2
167.21
MFCD00023871
YRTHJMQKDCXPAY-UHFFFAOYSA-N
3,3-tetramethyleneglutarimide, 8-azaspiro 4.5 decane-7,9-dione, azaspirodecanedione, 1,1-cyclopentanediacetimide, 8-aza-spiro 4.5 decane-7,9-dione, buspirone impurity k, dsstox_cid_31405, dsstox_rid_97291, dsstox_gsid_57616
136843
8-azaspiro[4.5]decane-7,9-dione
C1CCC2(C1)CC(=O)NC(=O)C2
Melting Point 153.0°C to 155.0°C
Color White
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Formula Weight 167.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylene glutarimide, 99%
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

RUO – Research Use Only