Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Tetramethylammonium Hydroxide Pentahydrate, ≥98%

Tetramethylammonium Hydroxide Pentahydrate is an ion pairing agent that is commonly used in analytical chemistry.

Supplier:  MP Biomedicals™ 0215211490

 View more versions of this product

Catalog No. ICN15211490


Add to Cart

Description

Description

  • Tetramethylammonium Hydroxide Pentahydrate is used in gas chromatography/mass spectrometry (GC/MS), capillary isotachophoresis and capillary zone electrophoresis, and liquid chromatography.
  • TM is also utilized in the synthesis of organic and inorganic compounds such as vinyl halides and tetramethylammonium dithiocarboxylates.
  • It is also used in the preparation of iron oxide nanoparticles for application as magnetic resonance imaging contrast agents.
  • It is an ion pairing agent that is commonly used in analytical chemistry.
  • It is also used as a developer or etchant in semiconductor and photoelectric industries. 
Specifications

Specifications

Tetramethylammonium Hydroxide Pentahydrate
Solid
White
MFCD00149566
O.O.O.O.O.[OH-].C[N+](C)(C)C
181.23
Store at 4°C.
500 g
C4H23NO6
MYXKPFMQWULLOH-UHFFFAOYSA-M
tetramethylazanium pentahydrate hydroxide
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.