Learn More
United Scientific™ Test Tube Clamp with Finger Grips
Test Tube Clamp for use with all standard test tube sizes.
$1.28 - $2.10
Specifications
Quantity | 1 Ea. |
---|---|
Type | Test Tube Clamp |
Color | Gold |
Jaw Type | Spring Loaded |
Description
- Self-closing clamp is designed to securely hold test tubes.
- They are constructed from brass wire and include finger grips for added comfort and ease of use.
Specifications
1 Ea. | |
Test Tube Clamp | |
Gold | |
Spring Loaded |