Learn More
Andwin Scientific TEST TB NS PRO VWR 17X100 M/CS
Supplier: Andwin Scientific 60818618
Polypropylene Culture Tube Without caps. Autoclavable. Size mm: 17x100. Volume: 14mL. Bulk packaging. Endotoxin-free., 1000/CS