Learn More
Hach Company Cyanuric Acid Test Kit, Model CY-3
Supplier: Hach Company 185102
A stabilizing agent for chlorine, cyanuric acid is used in swimming pool treatment. Cyanuric Acid Test Kit measures the turbidity formed in proportion to Cyanuric Acid concentration, after the reagent is added. Contains 100 tests.