Learn More
Hach Company Chlorine (Free & Total) Color Disc Test Kit, Model CN-66
Supplier: Hach Company 223101
This color disc test kit uses powder DPD reagent that reacts with chlorine more quickly than tablet-form DPD, giving more accurate results. Powder DPD also has a considerable advantage over orthotolidine, a hazardous substance sometimes used as a free and total chlorine test reagent. Hach Chlorine Test Kits are manufactured by Hach to ensure compliance with stringent quality control standards. Kit contains 100 tests.*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.