Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terumo™ Capiject™ Safety Lancets

High-quality, single-use lancets available in two formats: blades or needles

Supplier:  Terumo™ 200105

 View more versions of this product

Catalog No. 22-024-389


Add to Cart
This item is not returnable. View return policy

Description

Description

  • Automatic blade or needle retraction styles—the sharp edge is enclosed before and after use
  • Ready to use right out of the box
  • Convenient one-step activation for precise and consistent results
  • Color-coded sizes for quick identification of the right product

Specifications

Specifications

28 g
1.25 mm
Sterile
0.04 in.
Purple
200/Pk.
Single Blood Drop
For precise and consistent results
Automatic needle retraction
Safety Lancet
Safety and Handling

Safety and Handling

ShelfLife : 5 years

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit