Learn More
Eppendorf™ Temperature Sensor Device for Mastercycler
Rapid, easy to use, and suitable for use with Mastercycler PCR cyclers.
Specifications
Content And Storage | Requires Temperature Sensor |
---|---|
Description | Single Channel |
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
05-414-218
|
Eppendorf™
0056001000 |
Mastercycler, 96-well | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Measuring device without temperature sensor
- Sensor for 96 well and 384 well blocks, sold separately
- Simple, rapid verification and adjustment
- Automatic adjustment with Mastercycler software, version 2.10 or higher
- Can be third-party adjusted
Specifications
Requires Temperature Sensor | |
Single Channel |