Learn More
Bellco Glass, Inc. Optional temperature chart recorder for Roll-In Incubators. The chart recorder record up to 24 hours a day and 7 days a week. The unit is housed grey plastic case with a 6" viewing window. User can select temperature ranges (preset to 0-100C) & audible al
Supplier: Bellco Glass, Inc. 772835230
Optional temperature chart recorder for Roll-In Incubators. The chart recorder record up to 24 hours a day and 7 days a week. The unit is housed grey plastic case with a 6" viewing window. User can select temperature ranges (preset to 0-100C) & audible al