Learn More
Azzota Corp Temperature Probe For Heating Mantles, Length 9.5 Inch (240mm), Din Connector, 5 Pins
Supplier:  Azzota Corp HMTEMP
Encompass_Preferred
Temperature Probe for Heating Mantles..Length 9.5 inch (240mm), DIN connector, 5 pins
Catalog No. 50-161-2855
Due to product restrictions, please to purchase or view availability for this product.