Learn More
Wilmad Labglass TEMP CTRL POWERTROL 120V
Supplier: Wilmad Labglass LG8954100
Mantle Controller, PowrTrol. Volts: 120; Wattage: 1200; Amps: 10; Output Range: 5-100%. Solid-state proportional-voltage power control that provides precise manual control of heating mantles, tapes, cords and other resistive loads.