Learn More
Deltatrak Inc Temp Chart Recorder-29C-90D38C, 20 units per case
Supplier: Deltatrak Inc 16500
Records produced by these temperature chart recorders serve as corroborating evidence for insurance claims, quality assurance documentation and protection of your valuable products. Available models from 5-Day to 90-Day transit periods with three temperature ranges.DeltaTrak's In-Transit Temperature Chart Recorder, Model 16000-16502, is a highly reliable, time and temperature strip chart recorder record environmental conditions during storage and transportation of food, pharmaceuticals, chemicals, and other temp-sensitive commodities in refrigerated trucks, rail cars, ocean/air freight. Temperature Range Standard Model: -20F to 100F (-29C to 38C) High Range Model: 32F to 150F (0C to 65C) Low Range Model: -40F to 80F (-40C to 27C) Accuracy 2F (1C) or better Sensor Bi-metal coil Chart Pressure sensitive, total length 36 inches (91.4cm), dual scale F/C Case Lot Size/Weight 20 recorders per case/11lbs. (4.9kg) Cert/calibration CE mark by T.U.V., traceable to N.I.S.T.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.