Learn More
Andwin Scientific TEGOSEPT 1KG
Supplier: Andwin Scientific 20258
Tegosept is utilized to delay and decrease the speed of growth rates in the larval and pupal growth stages. This is sold in a quantity of 1 kilogram.