Learn More
Perkin Elmer US LLC PTFE Mira Mist Nebulizer, 0.2-2.5 mL/min
Supplier: Perkin Elmer US LLC N0777031
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The PTFE Mira Mist™ nebulizer produces excellent mist and can handle high level of particulates in sample. This Mira Mist nebulizer is for samples with high HF, Sulfuric acid, and some other solvents that would attack PEEK nebulizers.