Learn More
Cole-Parmer Cole-Parmer 1" NPT(F) T-fitting with 3/4" NPT(F), CPVC adapter installed in branch
Supplier: Cole-Parmer 2700380
This has a one inch NPT(F) connection on all three sides. The top has a 3/4 inch NPT(F) for fitting - reducing bushing installed.The self-cleaning electrode in 27003 series ranges from 5 to 80 degrees Celcius.