Learn More
DSI LLC Tee, 360um, quick mount, 150um bore, for PEEK and FS Tubing (for < 10 kpsi)
Supplier:  DSI LLC JRC360QTPK6
Encompass_Preferred
Tee, 360um, quick mount, 150um bore, for PEEK and FS Tubing (for < 10 kpsi)
Catalog No. 50-225-5033