Learn More
Cole-Parmer ESS GD0707-7000 Sampling Bags With Combination Valve, 1L
Supplier: Cole-Parmer 8656100
Gas sampling bags are constructed of 2-mil thick Tedlar with solid seam to keep gases contained. Unique, polypropylene combination valve features 3/16" OD on/off stem suitable for tubing connections and integral PTFE silicone septum for syringe sampling. Low permeability. Select from a variety of sizes to suit your needs. Case of 10.