Learn More
Techne™ PrimePRO QPCR DNA detection Kit, Herpes simplex Type 1 (HHV1) (without Mastermix)
Supplier:  Techne™ TKIT10091
Catalog No. TKIT10091
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Kits packed in a convenient foil pouch
  • Lyophilized (freeze dried)
  • No dry ice shipping required
  • Reduced shipping costs
  • Simple protocol, identical for each kit
  • 150 reactions per kit
  • Low cost per test
  • qPCR can be days faster, more accurate and more quantifiable than traditional microbiological methods
  • Suitable for any qPCR instrument with FAM/VIC detection
Specifications
Specifications
Lyophilized primer and probe mix, Lyophilized master mix, Lyophilized master mix resuspension buffer, Lyophilized ROX Ampule, High-copy DNA for standard curve, Internal extraction control DNA, Beta-actin endogenous control, RNAse/DNAse free water, Reverse transcriptase (RNA kits only)
Store at ambient temperature, stable for 18 months
any qPCR instrument with FAM/VIC detection
150
Microbial Detection
qPCR
QPCR DNA Detection Kit
150 Reactions
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
ShelfLife : 18 months