Learn More
National Optical Teaching Laboratory Biological Microscope with Camera
The Stellar Series teaching and laboratory biological microscopes adopt a new structural design, with a smoother appearance and stable performance.
Supplier: National Optical DSTELLAR 1 T4X5
Description
- The combination of a Stellar Series teaching biological microscope with 100X oil immersion lens and digital solution enables you to display and share live images.
- Discover the possibilities and benefits of adding an integrated tablet or microscope camera to your Pro Series microscope.
- Choose from integrated 8 inch Android-based tablet or WiFi enabled microscope camera.
Specifications
WiFi Enabled | |
Trinocular | |
4X, 10X, 40X, 100X | |
Achromatic | |
Camera |
WF 10X/18 mm | |
LED | |
Stellar | |
Biological Microscope |