Learn More
National Optical Teaching Laboratory Biological Microscope
The Stellar Series teaching and laboratory biological microscopes adopt a new structural design, with a smoother appearance and stable performance.
Supplier: National Optical DSTELLAR 1 B4
Description
- The Stellar Series teaching biological microscopes adopts a new structural design and updated appearance.
- This microscope comes with DIN Achromat objectives, mechanical stage, Coaxial adjustment, built in cord wrap, built in slide holder and LED illumination.
- Choose between binocular or trinocular heads with a choice of three objectives (4X, 10X, 40X) or four objectives which includes the 100X oil immersion lens.
Specifications
WF 10X/18 mm | |
LED | |
Stellar | |
Biological Microscope |
Binocular | |
4X, 10X, 40X, 100X | |
Achromatic |