Learn More
ADS Biotec Inc Triethylammonium Acetate Solution (TEAA) 2.0 M, 6 x 200 mL
Supplier: ADS Biotec Inc SP5890
Triethylammonium Acetate Solution is a superior ion-pairing reagent from ADS Biotec developed specifically for reversed phase HPLC separations of DNA and RNA.