Learn More
Cole-Parmer Environmental Express TDS100 StableWeigh TDS Vessels, 100/pk
Supplier: Cole-Parmer EW3521109
Pre-treated, pre-weighed, and disposable for your convenienceThis product complies with the following methods: SM 2540, ASTM D5907, BS EN 15216:2007Save time — cools faster, requires fewer drying and weighing cyclesDisposable to eliminate cross-contaminationThese weighing vessels reduce preparation time, perfect for busy labs analyzing a lot of samples. Each vessel complies with the following methods: SM 2540, ASTM D5907, BS EN 15216:2007. Each vessel is treated to conform with ASTM D5907 Standard Test Methods for Filterable Matter (Total Dissolved Solids) and Nonfilterable Matter (Total Suspended Solids) in Water, then individually printed with the final weight. The material is lightweight, inert to water or wastewater components, and heat resistant. Vessels cool quickly decreasing wait times between heating and weighing.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.