Learn More
SEDIMENT TESTING TC900 Sediment Test Card 500/case
Supplier:  SEDIMENT TESTING TC900 500/CS
Encompass
Filter: T-1313-1210 white dry laid, chembond solid color non-woven. Fiber: 100% 1.5D rayon. Binder: 100% acrylic latex. Weight: 2.48-3.03 oz. yd2. Thicknexx: 17.0-21.0 mils. Air Permeability: 135-235 Cfm/sq.ft. Wet Mullen Burst: 21.0-24.0. Meets USDA rating requiring a hold-back rate of 75 microns.
Catalog No. NC0358189