Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ TC Mps1 12
Potent and selective Mps1 kinase inhibitor
$251.72 - $1024.28
Specifications
Purity | >99% |
---|---|
Product Type | TC Mps1 12 |
Molecular Weight (g/mol) | 324.38 |
Inhibitors | TC Mps1 12 |
Description
Potent and selective monopolar spindle 1 (Mps1) inhibitor (IC50 = 6.4 nM). Selective for Mps1 over a panel of 95 kinases including JNK. Inhibits A549 lung carcinoma cell proliferation and attenuates A549 cell xenograft growth in mice. Orally active.Specifications
>99% | |
TC Mps1 12 | |
324.38 | |
TC Mps1 12 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title